Learn More
1-Heptylamine, 98+%, Thermo Scientific Chemicals
$63.29 - $383.01
Chemical Identifiers
CAS | 111-68-2 |
---|---|
Molecular Formula | C7H17N |
Molecular Weight (g/mol) | 115.22 |
MDL Number | MFCD00008244 |
InChI Key | WJYIASZWHGOTOU-UHFFFAOYSA-N |
Synonym | heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 |
PubChem CID | 8127 |
IUPAC Name | heptan-1-amine |
SMILES | CCCCCCCN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1792214
|
Thermo Scientific Chemicals
A1792214 |
25 g |
Each for $63.29
|
|
|||||
AAA1792218
|
Thermo Scientific Chemicals
A1792218 |
50 g |
Each for $93.88
|
|
|||||
AAA1792230
|
Thermo Scientific Chemicals
A1792230 |
250 g |
Each for $383.01
|
|
|||||
Description
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.
Solubility
Soluble in water 6791 mg/L @ 25°C.
Notes
Air sensitive. Store away from oxidizing agents, air, acids.
Chemical Identifiers
111-68-2 | |
115.22 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
8127 | |
CCCCCCCN |
C7H17N | |
MFCD00008244 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
heptan-1-amine |
Specifications
111-68-2 | |
0.775 | |
35°C (95°F) | |
C7H17N | |
CH3(CH2)6NH2 | |
25 g | |
1731688 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
heptan-1-amine | |
8127 | |
≥98% |
-23°C | |
154°C to 156°C | |
Amine-like | |
1.4245 | |
MFCD00008244 | |
UN2734 | |
Air Sensitive | |
Soluble in water 6791mg/L @ 25°C. | |
CCCCCCCN | |
115.22 | |
115.22 | |
1-Heptylamine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber : 203-895-8
RTECSNumber : MK0600000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only