Learn More
1-Hexanethiol, 96%, Thermo Scientific Chemicals
$66.29 - $476.32
Chemical Identifiers
CAS | 111-31-9 |
---|---|
Molecular Formula | C6H14S |
Molecular Weight (g/mol) | 118.24 |
MDL Number | MFCD00004909 |
InChI Key | PMBXCGGQNSVESQ-UHFFFAOYSA-N |
Synonym | 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan |
PubChem CID | 8106 |
IUPAC Name | hexane-1-thiol |
SMILES | CCCCCCS |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC215270050
|
Thermo Scientific Chemicals
215270050 |
5 mL | Glass bottle |
N/A
|
|
||||
AC215271000
|
Thermo Scientific Chemicals
215271000 |
100 mL | Glass bottle |
N/A
|
|
||||
AC215275000
|
Thermo Scientific Chemicals
215275000 |
500 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-31-9 | |
118.24 | |
PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
8106 | |
CCCCCCS |
C6H14S | |
MFCD00004909 | |
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
hexane-1-thiol |
Specifications
111-31-9 | |
100.0 | |
Colorless to Yellow | |
150.0°C to 154.0°C | |
Authentic | |
Glass bottle | |
1.4470 to 1.4510 | |
MFCD00004909 | |
01, 408 | |
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
hexane-1-thiol | |
8106 | |
96% | |
1-Hexanethiol |
95.0 | |
-81°C to 80°C | |
0.8400g/mL | |
30°C | |
95% min. (GC) | |
C6H14S | |
CH3(CH2)5SH | |
5 mL | |
0.84 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohols, ethyl ether, hydrocarbons | |
CCCCCCS | |
118.24 | |
118.24 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED
GHS Signal Word: Danger
EINECSNumber : 203-857-
RUO – Research Use Only