missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 354180250

 View more versions of this product

Catalog No. AC354180250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
Specifications

Specifications

1-Hexyl-3-methylimidazolium hexafluorophosphate
98.5
-73.5°C
1.3045g/mL
99%
C10H19F6N2P
MFCD03093296
1.3045
YPWSSSRXUOQNMQ-UHFFFAOYSA-N
1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate
2734175
312.24
Powder
304680-35-1
100.0
White
Authentic
Plastic bottle
1.4160 to 1.418
25 g
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
312.24
497 mPa.s (25°C)
99%

Chemical Identifiers

304680-35-1
312.24
YPWSSSRXUOQNMQ-UHFFFAOYSA-N
2734175
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1
C10H19F6N2P
MFCD03093296
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate
1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only