Learn More
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%, Thermo Scientific Chemicals
$433.78 - $433.78
Chemical Identifiers
CAS | 304680-35-1 |
---|---|
Molecular Formula | C10H19F6N2P |
Molecular Weight (g/mol) | 312.24 |
MDL Number | MFCD03093296 |
InChI Key | YPWSSSRXUOQNMQ-UHFFFAOYSA-N |
Synonym | 1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate |
PubChem CID | 2734175 |
IUPAC Name | 1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate |
SMILES | F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC354180250
|
Thermo Scientific Chemicals
354180250 |
25 g | Plastic bottle |
Each for $433.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquidChemical Identifiers
304680-35-1 | |
312.24 | |
YPWSSSRXUOQNMQ-UHFFFAOYSA-N | |
2734175 | |
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1 |
C10H19F6N2P | |
MFCD03093296 | |
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate | |
1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate |
Specifications
304680-35-1 | |
100.0 | |
White | |
Authentic | |
Plastic bottle | |
1.4160 to 1.418 | |
25 g | |
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate | |
F[P-](F)(F)(F)(F)F.CCCCCCN1C=C[N+](C)=C1 | |
312.24 | |
497 mPa.s (25°C) | |
99% | |
1-Hexyl-3-methylimidazolium hexafluorophosphate |
98.5 | |
-73.5°C | |
1.3045g/mL | |
99% | |
C10H19F6N2P | |
MFCD03093296 | |
1.3045 | |
YPWSSSRXUOQNMQ-UHFFFAOYSA-N | |
1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate | |
2734175 | |
312.24 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
RUO – Research Use Only