missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Hexyl-3-methylimidazolium Tetrafluoroborate 97.0+%, TCI America™
Supplier: TCI America H10995G
Specifications
1-Hexyl-3-methylimidazolium Tetrafluoroborate | |
C10H19BF4N2 | |
5 g | |
1-hexyl-3-methylimidazolium tetrafluoroborate, 1-n-hexyl-3-methylimidazolium tetrafluoroborate, hmimbf4, c6mim bf4, acmc-1ckkl, hmim bf4, hmim+bf4, dsstox_cid_29108, dsstox_rid_83327, dsstox_gsid_49252 | |
F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1 | |
254.08 | |
254.08 | |
Liquid |
244193-50-8 | |
MFCD03095459 | |
1760 | |
MFXLOVLEQJRXFP-UHFFFAOYSA-N | |
1-hexyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide | |
2734179 | |
≥97.0% (N) |
Chemical Identifiers
244193-50-8 | |
254.08 | |
MFXLOVLEQJRXFP-UHFFFAOYSA-N | |
2734179 | |
F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1 |
C10H19BF4N2 | |
MFCD03095459 | |
1-hexyl-3-methylimidazolium tetrafluoroborate, 1-n-hexyl-3-methylimidazolium tetrafluoroborate, hmimbf4, c6mim bf4, acmc-1ckkl, hmim bf4, hmim+bf4, dsstox_cid_29108, dsstox_rid_83327, dsstox_gsid_49252 | |
1-hexyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide |
Safety and Handling
TSCA : No