missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexyl-3-methylimidazolium Tetrafluoroborate 97.0+%, TCI America™
$96.94 - $287.06
Chemical Identifiers
| CAS | 244193-50-8 |
|---|---|
| Molecular Formula | C10H19BF4N2 |
| Molecular Weight (g/mol) | 254.08 |
| MDL Number | MFCD03095459 |
| InChI Key | MFXLOVLEQJRXFP-UHFFFAOYSA-N |
| Synonym | 1-hexyl-3-methylimidazolium tetrafluoroborate, 1-n-hexyl-3-methylimidazolium tetrafluoroborate, hmimbf4, c6mim bf4, acmc-1ckkl, hmim bf4, hmim+bf4, dsstox_cid_29108, dsstox_rid_83327, dsstox_gsid_49252 |
| PubChem CID | 2734179 |
| IUPAC Name | 1-hexyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide |
| SMILES | F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1 |
Chemical Identifiers
| 244193-50-8 | |
| 254.08 | |
| MFXLOVLEQJRXFP-UHFFFAOYSA-N | |
| 2734179 | |
| F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1 |
| C10H19BF4N2 | |
| MFCD03095459 | |
| 1-hexyl-3-methylimidazolium tetrafluoroborate, 1-n-hexyl-3-methylimidazolium tetrafluoroborate, hmimbf4, c6mim bf4, acmc-1ckkl, hmim bf4, hmim+bf4, dsstox_cid_29108, dsstox_rid_83327, dsstox_gsid_49252 | |
| 1-hexyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide |
Specifications
| 244193-50-8 | |
| MFCD03095459 | |
| 1760 | |
| MFXLOVLEQJRXFP-UHFFFAOYSA-N | |
| 1-hexyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide | |
| 2734179 | |
| ≥97.0% (N) | |
| 1-Hexyl-3-methylimidazolium Tetrafluoroborate |
| C10H19BF4N2 | |
| 5 g | |
| 1-hexyl-3-methylimidazolium tetrafluoroborate, 1-n-hexyl-3-methylimidazolium tetrafluoroborate, hmimbf4, c6mim bf4, acmc-1ckkl, hmim bf4, hmim+bf4, dsstox_cid_29108, dsstox_rid_83327, dsstox_gsid_49252 | |
| F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1 | |
| 254.08 | |
| 254.08 | |
| Liquid |
Safety and Handling
TSCA : No