missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Hydroxybenzotriazole Monohydrate 97.0+%, TCI America™
$348.40 - $348.40
Chemical Identifiers
CAS | 80029-43-2 |
---|---|
Molecular Formula | C6H7N3O2 |
Molecular Weight (g/mol) | 153.14 |
MDL Number | MFCD00065690 |
InChI Key | PJUPKRYGDFTMTM-UHFFFAOYSA-N |
Synonym | 1,2,3-Benzotriazol-1-ol, HOBt |
PubChem CID | 2796029 |
IUPAC Name | 1H-1,2,3-benzotriazol-1-ol hydrate |
SMILES | O.ON1N=NC2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H0468200G
|
TCI America
H0468200G |
200 g |
Each for $348.40
|
|
|||||
Chemical Identifiers
80029-43-2 | |
153.14 | |
PJUPKRYGDFTMTM-UHFFFAOYSA-N | |
2796029 | |
O.ON1N=NC2=CC=CC=C12 |
C6H7N3O2 | |
MFCD00065690 | |
1,2,3-Benzotriazol-1-ol, HOBt | |
1H-1,2,3-benzotriazol-1-ol hydrate |
Specifications
80029-43-2 | |
White | |
MFCD00065690 | |
3474 | |
PJUPKRYGDFTMTM-UHFFFAOYSA-N | |
1H-1,2,3-benzotriazol-1-ol hydrate | |
2796029 | |
≥97.0% (T) | |
1-Hydroxybenzotriazole Monohydrate |
160°C | |
C6H7N3O2 | |
200 g | |
1,2,3-Benzotriazol-1-ol, HOBt | |
O.ON1N=NC2=CC=CC=C12 | |
153.14 | |
135.13 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3579&(5)-3603
RTECSNumber : DM1288000
TSCA : No