missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Hydroxyethane-1,1-diphosphonic Acid (ca. 60% in Water, ca. 4.2mol/L), TCI America™
Supplier: TCI America H0587500G
Specifications
1-Hydroxyethane-1,1-diphosphonic Acid (ca. 60% in Water, ca. 4.2mol/L) | |
-40°C | |
105°C | |
MFCD00070585 | |
3265 | |
DBVJJBKOTRCVKF-UHFFFAOYSA-N | |
(1-hydroxy-1-phosphonoethyl)phosphonic acid | |
3305 | |
206.03 |
2809-21-4 | |
Yellow | |
C2H8O7P2 | |
500 g | |
etidronic acid, etidronate, hedp, 1-hydroxyethylidene-1,1-diphosphonic acid, ehdp, etidronsaeure, acetodiphosphonic acid, hydroxyethanediphosphonic acid, oxyethylidenediphosphonic acid, turpinal sl | |
CC(O)(P(=O)(O)O)P(=O)(O)O | |
206.027 | |
CHEBI:4907 | |
Liquid |
Chemical Identifiers
2809-21-4 | |
206.027 | |
DBVJJBKOTRCVKF-UHFFFAOYSA-N | |
3305 | |
(1-hydroxy-1-phosphonoethyl)phosphonic acid |
C2H8O7P2 | |
MFCD00070585 | |
etidronic acid, etidronate, hedp, 1-hydroxyethylidene-1,1-diphosphonic acid, ehdp, etidronsaeure, acetodiphosphonic acid, hydroxyethanediphosphonic acid, oxyethylidenediphosphonic acid, turpinal sl | |
CHEBI:4907 | |
CC(O)(P(=O)(O)O)P(=O)(O)O |
Safety and Handling
EINECSNumber : (2)-2936
RTECSNumber : SZ8562100
TSCA : Yes