missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Iodo-4-nitrobenzene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 122381000

 View more versions of this product

Catalog No. AC122381000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Iodo-4-nitrobenzene
636-98-6
100.0
Brown
100°C
98.5% min. (GC)
C6H4INO2
MFCD00007299
05, 253
Solubility in water: insoluble
C1=CC(=CC=C1[N+](=O)[O-])I
249.01
249.01
Powder
99%
98.5
171.0°C to 173.0°C
289.0°C (772.0 mmHg)
Authentic
Glass bottle
IC6H4NO2
100 g
benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene
SCCCFNJTCDSLCY-UHFFFAOYSA-N
1-iodo-4-nitrobenzene
12495
99%

Chemical Identifiers

636-98-6
249.01
SCCCFNJTCDSLCY-UHFFFAOYSA-N
12495
C1=CC(=CC=C1[N+](=O)[O-])I
C6H4INO2
MFCD00007299
benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene
1-iodo-4-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapor

GHS Signal Word: Warning

EINECSNumber : 211-272-7

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only