Learn More
1-Iodo-4-nitrobenzene, 99%
CAS: 636-98-6 | C6H4INO2 | 249.01 g/mol
$613.53 - $613.53
Chemical Identifiers
| CAS | 636-98-6 |
|---|---|
| Molecular Formula | C6H4INO2 |
| Molecular Weight (g/mol) | 249.01 |
| MDL Number | MFCD00007299 |
| InChI Key | SCCCFNJTCDSLCY-UHFFFAOYSA-N |
| Synonym | benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene |
| PubChem CID | 12495 |
| IUPAC Name | 1-iodo-4-nitrobenzene |
| SMILES | C1=CC(=CC=C1[N+](=O)[O-])I |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC122381000
|
Thermo Scientific Chemicals
122381000 |
100 g | Glass bottle |
Each for $613.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 636-98-6 | |
| 249.01 | |
| SCCCFNJTCDSLCY-UHFFFAOYSA-N | |
| 12495 | |
| C1=CC(=CC=C1[N+](=O)[O-])I |
| C6H4INO2 | |
| MFCD00007299 | |
| benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene | |
| 1-iodo-4-nitrobenzene |
Specifications
| 636-98-6 | |
| 100.0 | |
| Brown | |
| 100°C | |
| 98.5% min. (GC) | |
| C6H4INO2 | |
| MFCD00007299 | |
| 05, 253 | |
| Solubility in water: insoluble | |
| C1=CC(=CC=C1[N+](=O)[O-])I | |
| 249.01 | |
| 249.01 | |
| Powder |
| 98.5 | |
| 171.0°C to 173.0°C | |
| 289.0°C (772.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| IC6H4NO2 | |
| 100 g | |
| benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene | |
| SCCCFNJTCDSLCY-UHFFFAOYSA-N | |
| 1-iodo-4-nitrobenzene | |
| 12495 | |
| 99% | |
| 1-Iodo-4-nitrobenzene, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapor
GHS Signal Word: Warning
EINECSNumber : 211-272-7
TSCA : TSCA
RUO – Research Use Only