Learn More
1-Iodobutane, 98%, stabilized, Thermo Scientific Chemicals
$124.78 - $351.13
Chemical Identifiers
CAS | 542-69-8 |
---|---|
Molecular Formula | C4H9I |
Molecular Weight (g/mol) | 184.02 |
MDL Number | MFCD00001098 |
InChI Key | KMGBZBJJOKUPIA-UHFFFAOYSA-N |
Synonym | butyl iodide, butane, 1-iodo, n-butyl iodide, 1-iodo-butane, butane, iodo, iodobutane, 1-jodbutan, 1-jodbutan czech, unii-607a6cc46r, butyliodide |
PubChem CID | 10962 |
IUPAC Name | 1-iodobutane |
SMILES | CCCCI |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC173431000
|
Thermo Scientific Chemicals
173431000 |
100 mL | Glass bottle |
Each for $124.78
|
|
||||
AC173435000
|
Thermo Scientific Chemicals
173435000 |
500 mL | Glass bottle |
Each for $351.13
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
542-69-8 | |
184.02 | |
KMGBZBJJOKUPIA-UHFFFAOYSA-N | |
10962 | |
CCCCI |
C4H9I | |
MFCD00001098 | |
butyl iodide, butane, 1-iodo, n-butyl iodide, 1-iodo-butane, butane, iodo, iodobutane, 1-jodbutan, 1-jodbutan czech, unii-607a6cc46r, butyliodide | |
1-iodobutane |
Specifications
542-69-8,7440-50-8 | |
100.0 | |
Colorless to Yellow or Pink | |
130.0°C to 131.0°C | |
Authentic | |
Glass bottle | |
1.4975 to 1.4995 | |
MFCD00001098 | |
01, 123 | |
15, 1576 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol and ether | |
CCCCI | |
184.02 | |
10962 | |
98% | |
1-Iodobutane, 98% |
97.5 | |
-103.0°C | |
1.6100g/mL | |
33°C | |
97.5% min. (GC) | |
C4H9I | |
CH3(CH2)3I | |
100 mL | |
1.61 | |
butyl iodide, butane, 1-iodo, n-butyl iodide, 1-iodo-butane, butane, iodo, iodobutane, 1-jodbutan, 1-jodbutan czech, unii-607a6cc46r, butyliodide | |
KMGBZBJJOKUPIA-UHFFFAOYSA-N | |
1-iodobutane | |
copper | |
184.02 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protectio
GHS Signal Word: Danger
EINECSNumber : 208-824-4
RTECSNumber : EK4400000
TSCA : TSCA
RUO – Research Use Only