missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 174050500

 View more versions of this product

Catalog No. AC174050500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Iodopropane
107-08-4
100.0
Colorless to Yellow
101.0°C to 102.0°C
Authentic
Glass bottle
1.5030 to 1.5050
MFCD00001095
01, 113
15, 7974
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
CC(C)I
169.99
33643
99%
99%
98.5
-101.0°C
1.7400g/mL
44°C
1.5% max. (Sum of all by-products) (GC)
C3H7I
CH3CH2CH2I
50 mL
1.74
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
FMKOJHQHASLBPH-UHFFFAOYSA-N
1-iodopropane
copper
169.99
Liquid

Chemical Identifiers

107-08-4
169.99
FMKOJHQHASLBPH-UHFFFAOYSA-N
33643
CC(C)I
C3H7I
MFCD00001095
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
1-iodopropane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

GHS Signal Word: Warning

EINECSNumber : 203-460-2

RTECSNumber : TZ4100000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only