missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopropane, 99%, stabilized

CAS: 107-08-4 | C3H7I | 169.99 g/mol

$87.05 - $244.38

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
IUPAC Name 1-iodopropane
SMILES CC(C)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC174050500
SDS View Documents
Thermo Scientific Chemicals
174050500
50 mL Glass bottle
Each for $87.05
Only null left
 
AC174052500
SDS View Documents
Thermo Scientific Chemicals
174052500
250 mL Glass bottle
Each for $244.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-08-4
169.99
FMKOJHQHASLBPH-UHFFFAOYSA-N
33643
CC(C)I
C3H7I
MFCD00001095
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
1-iodopropane

Specifications

107-08-4
100.0
Colorless to Yellow
101.0°C to 102.0°C
Authentic
Glass bottle
1.5030 to 1.5050
MFCD00001095
01, 113
15, 7974
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
CC(C)I
169.99
33643
99%
1-Iodopropane, 99%
98.5
-101.0°C
1.7400g/mL
44°C
1.5% max. (Sum of all by-products) (GC)
C3H7I
CH3CH2CH2I
50 mL
1.74
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
FMKOJHQHASLBPH-UHFFFAOYSA-N
1-iodopropane
copper
169.99
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

GHS Signal Word: Warning

EINECSNumber : 203-460-2

RTECSNumber : TZ4100000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only