Learn More
1-Iodopropane, 99%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 174052500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Iodopropane | |
107-08-4 | |
100.0 | |
Colorless to Yellow | |
101.0°C to 102.0°C | |
Authentic | |
Glass bottle | |
1.5030 to 1.5050 | |
MFCD00001095 | |
01, 113 | |
15, 7974 | |
Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether | |
CC(C)I | |
169.99 | |
33643 | |
99% |
99% | |
98.5 | |
-101.0°C | |
1.7400g/mL | |
44°C | |
1.5% max. (Sum of all by-products) (GC) | |
C3H7I | |
CH3CH2CH2I | |
250 mL | |
1.74 | |
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes | |
FMKOJHQHASLBPH-UHFFFAOYSA-N | |
1-iodopropane | |
copper | |
169.99 | |
Liquid |
Chemical Identifiers
107-08-4 | |
169.99 | |
FMKOJHQHASLBPH-UHFFFAOYSA-N | |
33643 | |
CC(C)I |
C3H7I | |
MFCD00001095 | |
propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes | |
1-iodopropane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Warning
EINECSNumber : 203-460-2
RTECSNumber : TZ4100000
TSCA : TSCA
RUO – Research Use Only