Learn More
1-Methoxy-2-propanol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 041547AK
Description
1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Methoxy-2-propanol | |
-100°C | |
0.926 g/mL | |
119°C to 120°C | |
>99% | |
C4H10O2 | |
MFCD00004537 | |
UN3092 | |
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b | |
ARXJGSRGQADJSQ-UHFFFAOYNA-N | |
1-methoxypropan-2-ol | |
7900 | |
≥99% |
107-98-2 | |
Colorless | |
4 to 7 | |
35°C (95°F) | |
Alcohol-like | |
1.403 | |
250 mL | |
1731270 | |
Miscible with water,methanol and ether. | |
COCC(C)O | |
90.12 | |
90.12 | |
Liquid |
Chemical Identifiers
107-98-2 | |
90.12 | |
ARXJGSRGQADJSQ-UHFFFAOYNA-N | |
7900 | |
COCC(C)O |
C4H10O2 | |
MFCD00004537 | |
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b | |
1-methoxypropan-2-ol |
Safety and Handling
GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1-METHOXY-2-PROPANOL
EINECSNumber : 203-539-1
RTECSNumber : UB7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only