missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methoxy-2-propanol, 99+%, Thermo Scientific Chemicals

$50.21 - $657.67

Chemical Identifiers

CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41547AK
View Documents
Thermo Scientific Chemicals
041547AK
250 mL
Each for $50.21
 
AA41547K2
View Documents
Thermo Scientific Chemicals
041547K2
1 L
Each for $84.32
 
AA41547K7
View Documents
Thermo Scientific Chemicals
041547K7
4 L
Each for $214.97
 
AA41547M6
View Documents
Thermo Scientific Chemicals
041547M6
4 x 4 L
Each for $657.67
 
Description

Description

1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-98-2
90.12
ARXJGSRGQADJSQ-UHFFFAOYNA-N
7900
COCC(C)O
C4H10O2
MFCD00004537
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
1-methoxypropan-2-ol

Specifications

107-98-2
Colorless
4 to 7
35°C (95°F)
Alcohol-like
1.403
250 mL
1731270
Miscible with water,methanol and ether.
COCC(C)O
90.12
90.12
Liquid
-100°C
0.926 g/mL
119°C to 120°C
>99%
C4H10O2
MFCD00004537
UN3092
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
ARXJGSRGQADJSQ-UHFFFAOYNA-N
1-methoxypropan-2-ol
7900
≥99%
1-Methoxy-2-propanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1-METHOXY-2-PROPANOL

EINECSNumber : 203-539-1

RTECSNumber : UB7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only