missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methoxy-2-propanol, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041547K7

 View more versions of this product

Catalog No. AA41547K7


Explore available promotions

Description

Description

1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Methoxy-2-propanol
-100°C
0.926 g/mL
119°C to 120°C
>99%
C4H10O2
MFCD00004537
3092
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
ARXJGSRGQADJSQ-UHFFFAOYNA-N
1-methoxypropan-2-ol
7900
≥99%
107-98-2
Colorless
4 to 7
35°C (95°F)
Alcohol-like
1.403
4 L
1731270
Miscible with water,methanol and ether.
COCC(C)O
90.12
90.12
Liquid

Chemical Identifiers

107-98-2
90.12
ARXJGSRGQADJSQ-UHFFFAOYNA-N
7900
COCC(C)O
C4H10O2
MFCD00004537
1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
1-methoxypropan-2-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 1-METHOXY-2-PROPANOL

EINECSNumber : 203-539-1

RTECSNumber : UB7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only