Learn More
1-Methyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 156840500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Methyl-1-cyclohexene | |
591-49-1 | |
100.0 | |
0.8100g/mL | |
−3°C | |
98% min. (GC) | |
C7H12 | |
C6H9CH3 | |
05, 66 | |
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044 | |
CTMHWPIWNRWQEG-UHFFFAOYSA-N | |
1-methylcyclohexene | |
10ppm hydroquinone | |
96.17 | |
Liquid |
98+% | |
98.0 | |
Colorless | |
110.0°C to 111.0°C | |
Authentic | |
Glass bottle | |
1.4492 to 1.4512 | |
50 g | |
0.81 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and benzene | |
CC1=CCCCC1 | |
96.17 | |
11574 | |
98+% |
Chemical Identifiers
591-49-1 | |
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044 | |
1-methylcyclohexene |
CTMHWPIWNRWQEG-UHFFFAOYSA-N | |
11574 | |
CC1=CCCCC1 |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-718-0
TSCA : TSCA
RUO – Research Use Only