missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclohexene, 97%, stabilized

CAS: 591-49-1 | C7H12 | 96.17 g/mol

$116.03 - $447.95

Chemical Identifiers

CAS 591-49-1
InChI Key CTMHWPIWNRWQEG-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
PubChem CID 11574
IUPAC Name 1-methylcyclohexene
SMILES CC1=CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC156840100
SDS View Documents
Thermo Scientific Chemicals
156840100
10 g
Each for $116.03
Only null left
 
AC156840500
SDS View Documents
Thermo Scientific Chemicals
156840500
50 g
Each for $447.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

591-49-1
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
1-methylcyclohexene
CTMHWPIWNRWQEG-UHFFFAOYSA-N
11574
CC1=CCCCC1

Specifications

591-49-1
100.0
0.8100g/mL
−3°C
98% min. (GC)
C7H12
C6H9CH3
05, 66
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
CTMHWPIWNRWQEG-UHFFFAOYSA-N
1-methylcyclohexene
10ppm hydroquinone
96.17
Liquid
98.0
Colorless
110.0°C to 111.0°C
Authentic
Glass bottle
1.4492 to 1.4512
10 g
0.81
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and benzene
CC1=CCCCC1
96.17
11574
98+%
1-Methyl-1-cyclohexene, 98+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-718-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only