missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-1H-indazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 445130010
Specifications
1-Methyl-1H-indazole-4-boronic acid pinacol ester | |
96% min. (GC) | |
C14H19BN2O2 | |
1g | |
OZHFBNULPUBRJF-UHFFFAOYSA-N | |
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole | |
45480187 | |
97% |
885698-94-2 | |
Glass Bottle | |
MFCD10700158 | |
1-methyl-1h-indazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methylindazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methyl-1h-indazole-4-boronic acid, pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole, 1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1 | |
258.13 | |
258.13 |
Chemical Identifiers
885698-94-2 | |
258.13 | |
OZHFBNULPUBRJF-UHFFFAOYSA-N | |
45480187 | |
CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1 |
C14H19BN2O2 | |
MFCD10700158 | |
1-methyl-1h-indazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methylindazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methyl-1h-indazole-4-boronic acid, pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole, 1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only