missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-indazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Chemical Identifiers

CAS 885698-94-2
Molecular Formula C14H19BN2O2
Molecular Weight (g/mol) 258.13
MDL Number MFCD10700158
InChI Key OZHFBNULPUBRJF-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methylindazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methyl-1h-indazole-4-boronic acid, pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole, 1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 45480187
IUPAC Name 1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
SMILES CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445130010
SDS View Documents
Thermo Scientific Chemicals
445130010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

885698-94-2
258.13
OZHFBNULPUBRJF-UHFFFAOYSA-N
45480187
CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1
C14H19BN2O2
MFCD10700158
1-methyl-1h-indazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methylindazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methyl-1h-indazole-4-boronic acid, pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole, 1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole

Specifications

885698-94-2
Glass Bottle
MFCD10700158
1-methyl-1h-indazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methylindazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methyl-1h-indazole-4-boronic acid, pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole, 1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1
258.13
258.13
1-Methyl-1H-indazole-4-boronic acid pinacol ester
96% min. (GC)
C14H19BN2O2
1g
OZHFBNULPUBRJF-UHFFFAOYSA-N
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
45480187
97%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only