missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylimidazole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 126991000

 View more versions of this product

Catalog No. AC126991000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Methylimidazole
616-47-7
100.0
Colorless to Yellow
198.0°C
Authentic
Glass bottle
1.4955 to 1.4975
100 g
05,447
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1-methylimidazole
1390
82.12
Liquid
99%
98.5
-2.0°C
1.0300g/mL
92°C
98.5% min. (GC)
C4H6N2
MFCD00005292
23, 46
1.03
Solubility in water: 145g/L (25°C). Other solubilities: soluble in organic solvents
CN1C=CN=C1
82.11
CHEBI:113454
99%

Chemical Identifiers

616-47-7
82.11
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1390
1-methylimidazole
C4H6N2
MFCD00005292
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
CHEBI:113454
CN1C=CN=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 210-484-7

RTECSNumber : NI7000000

TSCA : TSCA

Recommended Storage : this product turns yellow during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only