missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methylimidazole, 99%, Thermo Scientific Chemicals

$77.84 - $1,661.99

Chemical Identifiers

CAS 616-47-7
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005292
InChI Key MCTWTZJPVLRJOU-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
PubChem CID 1390
ChEBI CHEBI:113454
IUPAC Name 1-methylimidazole
SMILES CN1C=CN=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC126991000
SDS View Documents
Thermo Scientific Chemicals
126991000
100 g Glass bottle N/A
 
AC126995000
SDS View Documents
Thermo Scientific Chemicals
126995000
500 g Glass bottle N/A
 
AC126990050
SDS View Documents
Thermo Scientific Chemicals
126990050
5 kg Plastic drum N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

616-47-7
82.11
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1390
1-methylimidazole
C4H6N2
MFCD00005292
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
CHEBI:113454
CN1C=CN=C1

Specifications

616-47-7
100.0
Colorless to Yellow
198.0°C
Authentic
Glass bottle
1.4955 to 1.4975
100 g
05,447
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1-methylimidazole
1390
82.12
Liquid
98.5
-2.0°C
1.0300g/mL
92°C
98.5% min. (GC)
C4H6N2
MFCD00005292
23, 46
1.03
Solubility in water: 145g/L (25°C). Other solubilities: soluble in organic solvents
CN1C=CN=C1
82.11
CHEBI:113454
99%
1-Methylimidazole, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 210-484-7

RTECSNumber : NI7000000

TSCA : TSCA

Recommended Storage : this product turns yellow during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only