Learn More
1-n-Hexyl-3-methylimidazolium chloride, 98%
CAS: 171058-17-6 | C10H19N2 | 167.28 g/mol
Supplier: Thermo Scientific Chemicals H2717806
Description
1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
Specifications
| 1-n-Hexyl-3-methylimidazolium chloride | |
| C10H19N2 | |
| MFCD03093289 | |
| 1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f | |
| RVEJOWGVUQQIIZ-UHFFFAOYSA-N | |
| 1-hexyl-3-methylimidazol-3-ium;chloride | |
| 2734163 | |
| 202.72 |
| 171058-17-6 | |
| 1.5165 | |
| 5 g | |
| Insoluble in water. | |
| CCCCCC[N+]1=CN(C)C=C1 | |
| 167.28 | |
| CHEBI:61340 | |
| 98% |
Chemical Identifiers
| 171058-17-6 | |
| 167.28 | |
| RVEJOWGVUQQIIZ-UHFFFAOYSA-N | |
| 2734163 | |
| 1-hexyl-3-methylimidazol-3-ium;chloride |
| C10H19N2 | |
| MFCD03093289 | |
| 1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f | |
| CHEBI:61340 | |
| CCCCCC[N+]1=CN(C)C=C1 |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only