Learn More
1-n-Hexyl-3-methylimidazolium chloride, 98%, Thermo Scientific Chemicals
$71.22 - $321.41
Chemical Identifiers
CAS | 171058-17-6 |
---|---|
Molecular Formula | C10H19N2 |
Molecular Weight (g/mol) | 167.28 |
MDL Number | MFCD03093289 |
InChI Key | RVEJOWGVUQQIIZ-UHFFFAOYSA-N |
Synonym | 1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f |
PubChem CID | 2734163 |
ChEBI | CHEBI:61340 |
IUPAC Name | 1-hexyl-3-methylimidazol-3-ium;chloride |
SMILES | CCCCCC[N+]1=CN(C)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH2717806
|
Thermo Scientific Chemicals
H2717806 |
5 g |
Each for $71.22
|
|
||||||
AAH2717818
|
Thermo Scientific Chemicals
H2717818 |
50 g |
Each for $321.41
|
|
||||||
Description
1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
Chemical Identifiers
171058-17-6 | |
167.28 | |
RVEJOWGVUQQIIZ-UHFFFAOYSA-N | |
2734163 | |
1-hexyl-3-methylimidazol-3-ium;chloride |
C10H19N2 | |
MFCD03093289 | |
1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f | |
CHEBI:61340 | |
CCCCCC[N+]1=CN(C)C=C1 |
Specifications
171058-17-6 | |
1.5165 | |
5 g | |
Insoluble in water. | |
CCCCCC[N+]1=CN(C)C=C1 | |
167.28 | |
CHEBI:61340 | |
98% |
C10H19N2 | |
MFCD03093289 | |
1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f | |
RVEJOWGVUQQIIZ-UHFFFAOYSA-N | |
1-hexyl-3-methylimidazol-3-ium;chloride | |
2734163 | |
202.72 | |
1-n-Hexyl-3-methylimidazolium chloride |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only