Learn More
1-Naphthoic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 128180250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Naphthoic acid | |
86-55-5 | |
100.0 | |
Authentic | |
Plastic bottle | |
C10H7CO2H | |
25g | |
15, 6466 | |
Solubility in water: slightly soluble in hot water. Other solubilities: freely soluble in hot alcohol and ether | |
C1=CC=C2C(=C1)C=CC=C2C(=O)O | |
172.18 | |
CHEBI:36466 | |
98% |
98% | |
97.5 | |
300.0°C | |
97.5% min. (HPLC) | |
C11H8O2 | |
MFCD00004007 | |
09, 647 | |
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh | |
LNETULKMXZVUST-UHFFFAOYSA-N | |
naphthalene-1-carboxylic acid | |
6847 | |
172.18 |
Chemical Identifiers
86-55-5 | |
172.18 | |
LNETULKMXZVUST-UHFFFAOYSA-N | |
6847 | |
naphthalene-1-carboxylic acid |
C11H8O2 | |
MFCD00004007 | |
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh | |
CHEBI:36466 | |
C1=CC=C2C(=C1)C=CC=C2C(=O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 201-681-9
RTECSNumber : QL0960000
TSCA : TSCA
RUO â Research Use Only