missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthoic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 86-55-5
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD00004007
InChI Key LNETULKMXZVUST-UHFFFAOYSA-N
Synonym 1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
PubChem CID 6847
ChEBI CHEBI:36466
IUPAC Name naphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC=C2C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128180250
SDS View Documents
Thermo Scientific Chemicals
128180250
25g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC128181000
SDS View Documents
Thermo Scientific Chemicals
128181000
100g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

86-55-5
172.18
LNETULKMXZVUST-UHFFFAOYSA-N
6847
naphthalene-1-carboxylic acid
C11H8O2
MFCD00004007
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
CHEBI:36466
C1=CC=C2C(=C1)C=CC=C2C(=O)O

Specifications

86-55-5
100.0
Authentic
Plastic bottle
C10H7CO2H
25g
15, 6466
Solubility in water: slightly soluble in hot water. Other solubilities: freely soluble in hot alcohol and ether
C1=CC=C2C(=C1)C=CC=C2C(=O)O
172.18
CHEBI:36466
98%
97.5
300.0°C
97.5% min. (HPLC)
C11H8O2
MFCD00004007
09, 647
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
LNETULKMXZVUST-UHFFFAOYSA-N
naphthalene-1-carboxylic acid
6847
172.18
1-Naphthoic acid, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 201-681-9

RTECSNumber : QL0960000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only