missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 128181000

 View more versions of this product

Catalog No. AC128181000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Naphthoic acid
86-55-5
100.0
Authentic
Plastic bottle
C10H7CO2H
100g
15, 6466
Solubility in water: slightly soluble in hot water. Other solubilities: freely soluble in hot alcohol and ether
C1=CC=C2C(=C1)C=CC=C2C(=O)O
172.18
CHEBI:36466
98%
98%
97.5
300.0°C
97.5% min. (HPLC)
C11H8O2
MFCD00004007
09, 647
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
LNETULKMXZVUST-UHFFFAOYSA-N
naphthalene-1-carboxylic acid
6847
172.18

Chemical Identifiers

86-55-5
172.18
LNETULKMXZVUST-UHFFFAOYSA-N
6847
naphthalene-1-carboxylic acid
C11H8O2
MFCD00004007
1-naphthoic acid, 1-naphthalenecarboxylic acid, 1-carboxynaphthalene, naphthoic acid, alpha-naphthoic acid, naphthalenecarboxylic acid, carboxynaphthalene, alpha-naphthylcarboxylic acid, naphthalene-alpha-carboxylic acid, unii-2niv4o66bh
CHEBI:36466
C1=CC=C2C(=C1)C=CC=C2C(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 201-681-9

RTECSNumber : QL0960000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only