missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Naphthyl butyrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 334950050

 View more versions of this product

Catalog No. AC334950050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Naphthyl butyrate
3121-70-8
100.0
1.0900g/mL
62°C
97.5% min. (GC)
C14H14O2
5 g
1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
naphthalen-1-yl butanoate
76571
98%
98%
97.5
Yellow
125°C to 130°C (1.0 mmHg)
Authentic
Glass Bottle
MFCD00003923
1.09
Solubility in water: immiscible in water
CCCC(=O)OC1=CC=CC2=CC=CC=C21
214.26
214.26
Liquid

Chemical Identifiers

3121-70-8
214.26
XIVJNRXPRQKFRZ-UHFFFAOYSA-N
76571
CCCC(=O)OC1=CC=CC2=CC=CC=C21
C14H14O2
MFCD00003923
1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik
naphthalen-1-yl butanoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-500-7

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only