Learn More
1-Naphthyl isocyanate, 99%, Thermo Scientific Chemicals
$108.25 - $359.47
Chemical Identifiers
CAS | 86-84-0 |
---|---|
InChI Key | BDQNKCYCTYYMAA-UHFFFAOYSA-N |
Synonym | 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester |
PubChem CID | 66589 |
IUPAC Name | 1-isocyanatonaphthalene |
SMILES | C1=CC=C2C(=C1)C=CC=C2N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC415410050
|
Thermo Scientific Chemicals
415410050 |
5 g |
Each for $108.25
|
|
|||||
AC415410250
|
Thermo Scientific Chemicals
415410250 |
25 g |
Each for $359.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
86-84-0 | |
1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester | |
1-isocyanatonaphthalene |
BDQNKCYCTYYMAA-UHFFFAOYSA-N | |
66589 | |
C1=CC=C2C(=C1)C=CC=C2N=C=O |
Specifications
86-84-0 | |
100.0 | |
1.1800g/mL | |
135°C | |
Glass bottle | |
1.6324 to 1.6344 | |
1.18 | |
1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester | |
BDQNKCYCTYYMAA-UHFFFAOYSA-N | |
1-isocyanatonaphthalene | |
66589 | |
99% |
98.5 | |
4°C | |
269°C to 270°C | |
99% | |
C11H7NO | |
5 g | |
15, 6492 | |
Solubility in water: decomposes. Other solubilities: soluble in toluene,chloroform,ether,decomposes in alcohols | |
C1=CC=C2C(=C1)C=CC=C2N=C=O | |
169.18 | |
169.18 | |
1-Naphthyl isocyanate |
Safety and Handling
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-703-7
RUO – Research Use Only