missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 151530250

 View more versions of this product

Catalog No. AC151530250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Nitroso-2-naphthol
131-91-9
100.0
Brown
98%
C10H7NO2
MFCD00003884
07, 712
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
1-nitrosonaphthalen-2-ol
8580
98%
98%
97.5
104.0°C to 108.0°C
Authentic
Glass bottle
ONC10H6OH
25 g
15, 6728
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene,carbon,disulfide,caustic alkali solutions and glacial,acetic acid,slightly soluble in cold petroleum ether
C1=CC=C2C(=C1)C=CC(=C2N=O)O
173.17
173.17
Powder

Chemical Identifiers

131-91-9
173.17
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
8580
C1=CC=C2C(=C1)C=CC(=C2N=O)O
C10H7NO2
MFCD00003884
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
1-nitrosonaphthalen-2-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a posit

GHS Signal Word: Warning

EINECSNumber : 205-043-0

RTECSNumber : QL4725000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only