missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Nitroso-2-naphthol, 98%, Thermo Scientific Chemicals

$93.48 - $1439.16

Chemical Identifiers

CAS 131-91-9
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00003884
InChI Key YXAOOTNFFAQIPZ-UHFFFAOYSA-N
Synonym 1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
PubChem CID 8580
IUPAC Name 1-nitrosonaphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2N=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151530250
SDS View Documents
Thermo Scientific Chemicals
151530250
25 g Glass bottle
Each for $93.48
 
AC151535000
SDS View Documents
Thermo Scientific Chemicals
151535000
500 g Glass bottle
Each for $1,439.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

131-91-9
173.17
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
8580
C1=CC=C2C(=C1)C=CC(=C2N=O)O
C10H7NO2
MFCD00003884
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
1-nitrosonaphthalen-2-ol

Specifications

131-91-9
100.0
Brown
98%
C10H7NO2
MFCD00003884
07, 712
1-nitroso-2-naphthol, 2-naphthalenol, 1-nitroso, nitroso-beta-naphthol, 2-naphthol, 1-nitroso, 1-nitroso-2-naftol, zelen moridlova 4, zelen moridlova 4 czech, alpha-nitroso-beta-naphthol, 1-nitroso-2-naftol czech, unii-757i55u2qx
YXAOOTNFFAQIPZ-UHFFFAOYSA-N
1-nitrosonaphthalen-2-ol
8580
98%
1-Nitroso-2-naphthol, 98%
97.5
104.0°C to 108.0°C
Authentic
Glass bottle
ONC10H6OH
25 g
15, 6728
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene,carbon,disulfide,caustic alkali solutions and glacial,acetic acid,slightly soluble in cold petroleum ether
C1=CC=C2C(=C1)C=CC(=C2N=O)O
173.17
173.17
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a posit

GHS Signal Word: Warning

EINECSNumber : 205-043-0

RTECSNumber : QL4725000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only