missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octadecylamine hydrochloride, Thermo Scientific Chemicals

$87.19 - $291.60

Chemical Identifiers

CAS 1838-08-0
Molecular Formula C18H40ClN
Molecular Weight (g/mol) 305.975
MDL Number MFCD00042018
InChI Key RNYJXPUAFDFIQJ-UHFFFAOYSA-N
Synonym octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
PubChem CID 519552
IUPAC Name octadecan-1-amine;hydrochloride
SMILES CCCCCCCCCCCCCCCCCCN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4137806
View Documents
Thermo Scientific Chemicals
04137806
5 g
Each for $87.19
 
AA4137814
View Documents
Thermo Scientific Chemicals
04137814
25 g
Each for $291.60
 
Description

Description

1-Octadecylamine hydrochloride is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1838-08-0
305.975
RNYJXPUAFDFIQJ-UHFFFAOYSA-N
519552
CCCCCCCCCCCCCCCCCCN.Cl
C18H40ClN
MFCD00042018
octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
octadecan-1-amine;hydrochloride

Specifications

1838-08-0
C18H40ClN
5 g
Soluble in hot methanol.
CCCCCCCCCCCCCCCCCCN.Cl
305.975
305.97
1-Octadecylamine hydrochloride
160°C
MFCD00042018
octadecylamine hydrochloride, stearylamine hydrochloride, 1-octadecylamine hydrochloride, stearamine hydrochloride, unii-3v3v50q0sj, 1-octadecanamine, hydrochloride, octadecylamine, hydrochloride, acmc-1bnqe, octadecyl-ammonium chloride, c18h39n.hcl
RNYJXPUAFDFIQJ-UHFFFAOYSA-N
octadecan-1-amine;hydrochloride
519552
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 217-410-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only