missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octanesulfonic Acid, Sodium Salt Anhydrous, HPLC Grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 416361000

 View more versions of this product

Catalog No. AC416361000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
Specifications

Specifications

1-Octanesulfonic Acid, Sodium Salt
(2.7 g/50 ml water) (Acidified with ACS grade Phosphoric Acid to pH 3), 0.04 max. at 254nm, 0.04 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm
White
Authentic
CH3(CH2)7SO3Na
100 g
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium octane-1-sulfonate
23669624
HPLC
5324-84-5
>300°C
1.5% max. (1 g, 110°C, 75 min)
C8H17NaO3S
MFCD00007544
04, III, 25
Solubility in water: freely soluble. Other solubilities: 4g/L in methanol
[Na+].CCCCCCCCS([O-])(=O)=O
216.27
216.28
Crystalline Solid

Chemical Identifiers

5324-84-5
216.27
HRQDCDQDOPSGBR-UHFFFAOYSA-M
23669624
[Na+].CCCCCCCCS([O-])(=O)=O
C8H17NaO3S
MFCD00007544
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
sodium octane-1-sulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 226-195-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only