missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octanesulfonic acid, sodium salt monohydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 206140250

 View more versions of this product

Catalog No. AC206140250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Octanesulfonic acid, sodium salt monohydrate
>300.0°C
Authentic
Glass bottle
CH3(CH2)7SO3Na·H2O
25 g
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
MBURIAHQXJQKRE-UHFFFAOYSA-M
sodium;octane-1-sulfonate;hydrate
23666339
98%
207596-29-0
White
98%
C8H19NaO4S
MFCD00149551
04,III,25
Solubility in water: soluble. Other solubilities: 4g/l methanol
O.[Na+].CCCCCCCCS([O-])(=O)=O
234.29
234.3
Fine Powder

Chemical Identifiers

207596-29-0
234.29
MBURIAHQXJQKRE-UHFFFAOYSA-M
23666339
O.[Na+].CCCCCCCCS([O-])(=O)=O
C8H19NaO4S
MFCD00149551
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
sodium;octane-1-sulfonate;hydrate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only