missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octanesulfonic acid, sodium salt monohydrate, 98%, Thermo Scientific Chemicals

$325.87 - $1,067.94

Chemical Identifiers

CAS 207596-29-0
Molecular Formula C8H19NaO4S
Molecular Weight (g/mol) 234.29
MDL Number MFCD00149551
InChI Key MBURIAHQXJQKRE-UHFFFAOYSA-M
Synonym sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
PubChem CID 23666339
IUPAC Name sodium;octane-1-sulfonate;hydrate
SMILES O.[Na+].CCCCCCCCS([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC206140250
SDS View Documents
Thermo Scientific Chemicals
206140250
25 g Glass bottle N/A
 
AC206141000
SDS View Documents
Thermo Scientific Chemicals
206141000
100 g Glass Bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207596-29-0
234.29
MBURIAHQXJQKRE-UHFFFAOYSA-M
23666339
O.[Na+].CCCCCCCCS([O-])(=O)=O
C8H19NaO4S
MFCD00149551
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
sodium;octane-1-sulfonate;hydrate

Specifications

207596-29-0
White
98%
C8H19NaO4S
MFCD00149551
04,III,25
Solubility in water: soluble. Other solubilities: 4g/l methanol
O.[Na+].CCCCCCCCS([O-])(=O)=O
234.29
234.3
Fine Powder
>300.0°C
Authentic
Glass bottle
CH3(CH2)7SO3Na·H2O
25 g
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
MBURIAHQXJQKRE-UHFFFAOYSA-M
sodium;octane-1-sulfonate;hydrate
23666339
98%
1-Octanesulfonic acid, sodium salt monohydrate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only