missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Octanesulfonic Acid, Sodium salt, monohydrate, HPLC grade, 99% min., MilliporeSigma™
Supplier: Technidata OX01013
Specifications
1-Octanesulfonic Acid, Sodium Salt, Monohydrate | |
207596-29-0 | |
Glass Bottle | |
CH3(CH2)7SO3Na · H2O | |
100 g | |
MBURIAHQXJQKRE-UHFFFAOYSA-M | |
sodium hydrate octane-1-sulfonate | |
23666339 | |
HPLC |
For analysis of HPLC | |
99% min. | |
C8H19NaO4S | |
MFCD00149551 | |
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate | |
O.[Na+].CCCCCCCCS([O-])(=O)=O | |
234.29 | |
234.29 |
Chemical Identifiers
207596-29-0 | |
234.29 | |
MBURIAHQXJQKRE-UHFFFAOYSA-M | |
23666339 | |
O.[Na+].CCCCCCCCS([O-])(=O)=O |
C8H19NaO4S | |
MFCD00149551 | |
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate | |
sodium hydrate octane-1-sulfonate |
Safety and Handling
Recommended Storage : Room Temperature