missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanesulfonic Acid, Sodium salt, monohydrate, HPLC grade, 99% min., MilliporeSigma™
$378.03 - $1239.01
Chemical Identifiers
| CAS | 207596-29-0 |
|---|---|
| Molecular Formula | C8H19NaO4S |
| Molecular Weight (g/mol) | 234.29 |
| MDL Number | MFCD00149551 |
| InChI Key | MBURIAHQXJQKRE-UHFFFAOYSA-M |
| Synonym | sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate |
| PubChem CID | 23666339 |
| IUPAC Name | sodium hydrate octane-1-sulfonate |
| SMILES | O.[Na+].CCCCCCCCS([O-])(=O)=O |
Chemical Identifiers
| 207596-29-0 | |
| 234.29 | |
| MBURIAHQXJQKRE-UHFFFAOYSA-M | |
| 23666339 | |
| O.[Na+].CCCCCCCCS([O-])(=O)=O |
| C8H19NaO4S | |
| MFCD00149551 | |
| sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate | |
| sodium hydrate octane-1-sulfonate |
Specifications
| 207596-29-0 | |
| C8H19NaO4S | |
| MFCD00149551 | |
| sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate | |
| O.[Na+].CCCCCCCCS([O-])(=O)=O | |
| 234.29 | |
| 234.29 | |
| 1-Octanesulfonic Acid, Sodium salt, monohydrate, For analysis of HPLC |
| 99% min. | |
| CH3(CH2)7SO3Na · H2O | |
| 25 g | |
| MBURIAHQXJQKRE-UHFFFAOYSA-M | |
| sodium hydrate octane-1-sulfonate | |
| 23666339 | |
| HPLC |
Safety and Handling
Recommended Storage : Room Temperature