missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, 99%, anhydrous, AcroSeal™

CAS: 111-87-5 | C8H18O | 130.23 g/mol

Supplier:  Thermo Scientific Chemicals 434580010

 View more versions of this product

Catalog No. AC434580010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Octanol
98.5
-16.0°C
0.8240g/mL
81°C
98.5% min. (GC)
C8H18O
CH3(CH2)7OH
1 L
0.824
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
KBPLFHHGFOOTCA-UHFFFAOYSA-N
octan-1-ol
957
CHEBI:16188
99%
111-87-5
100.0
Colorless to Yellow
195.0°C
Authentic
AcroSeal™ Glass Bottle
1.4280 to 1.4310
MFCD00002988
0.004 % max.
15,684
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,and mineral oil,soluble in propylene glycol,insoluble in glycerol
CCCCCCCCO
130.23
9 mPa.s (20°C)
130.23
Liquid

Chemical Identifiers

111-87-5
130.23
KBPLFHHGFOOTCA-UHFFFAOYSA-N
957
octan-1-ol
C8H18O
MFCD00002988
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
CHEBI:16188
CCCCCCCCO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas

GHS Signal Word: Warning

EINECSNumber : 203-917-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only