missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octanol, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

$92.44 - $374.77

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434581000
SDS View Documents
Thermo Scientific Chemicals
434581000
100 mL AcroSeal™ Glass Bottle
Each for $92.44
 
AC434580010
SDS View Documents
Thermo Scientific Chemicals
434580010
1 L AcroSeal™ Glass Bottle
Each for $374.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-87-5
130.23
KBPLFHHGFOOTCA-UHFFFAOYSA-N
957
octan-1-ol
C8H18O
MFCD00002988
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
CHEBI:16188
CCCCCCCCO

Specifications

111-87-5
100.0
Colorless to Yellow
195.0°C
Authentic
AcroSeal™ Glass Bottle
1.4280 to 1.4310
MFCD00002988
0.004 % max.
15,684
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,and mineral oil,soluble in propylene glycol,insoluble in glycerol
CCCCCCCCO
130.23
9 mPa.s (20°C)
130.23
Liquid
98.5
-16.0°C
0.8240g/mL
81°C
98.5% min. (GC)
C8H18O
CH3(CH2)7OH
100 mL
0.824
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
KBPLFHHGFOOTCA-UHFFFAOYSA-N
octan-1-ol
957
CHEBI:16188
99%
1-Octanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas

GHS Signal Word: Warning

EINECSNumber : 203-917-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only