missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1265614

 View more versions of this product

Catalog No. AAA1265614


Explore available promotions

Description

Description

1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

Solubility
Soluble in 5% ethanol, water (partly), acetone, chloroform, and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications

Specifications

1-Phenyl-1H-tetrazole-5-thiol
∼145°C (decomposition)
Odorless
MFCD00003129
UN1325
1-phenyl-1h-tetrazole-5-thiol, 1-phenyltetrazole-5-thiol, 1-phenyl-5-mercaptotetrazole, 5-mercapto-1-phenyltetrazole, 1-phenyltetrazoline-5-thione, mercaptophenyltetrazole, phenylmercaptotetrazole, 1-phenyl-5-tetrazolethione, 5-mercapto-1-phenyltetrazol, 1-phenyltetrazole-thiol
GGZHVNZHFYCSEV-UHFFFAOYSA-N
1-phenyl-2H-tetrazole-5-thione
690730
178.22
86-93-1
138°C (280°F)
C7H6N4S
25 g
139068
Soluble in 5% ethanol,water (partly),acetone,chloroform,and methanol.
S=C1N=NNN1C1=CC=CC=C1
178.21
CHEBI:79916
99%

Chemical Identifiers

86-93-1
178.21
GGZHVNZHFYCSEV-UHFFFAOYSA-N
690730
S=C1N=NNN1C1=CC=CC=C1
C7H6N4S
MFCD00003129
1-phenyl-1h-tetrazole-5-thiol, 1-phenyltetrazole-5-thiol, 1-phenyl-5-mercaptotetrazole, 5-mercapto-1-phenyltetrazole, 1-phenyltetrazoline-5-thione, mercaptophenyltetrazole, phenylmercaptotetrazole, 1-phenyl-5-tetrazolethione, 5-mercapto-1-phenyltetrazol, 1-phenyltetrazole-thiol
CHEBI:79916
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P370+P378q-P501c

H228-H317-H319

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 201-710-5

RTECSNumber : XF7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only