missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1H-tetrazole-5-thiol, 99%, Thermo Scientific Chemicals

$77.82 - $1219.26

Chemical Identifiers

CAS 86-93-1
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00003129
InChI Key GGZHVNZHFYCSEV-UHFFFAOYSA-N
Synonym 1-phenyl-1h-tetrazole-5-thiol, 1-phenyltetrazole-5-thiol, 1-phenyl-5-mercaptotetrazole, 5-mercapto-1-phenyltetrazole, 1-phenyltetrazoline-5-thione, mercaptophenyltetrazole, phenylmercaptotetrazole, 1-phenyl-5-tetrazolethione, 5-mercapto-1-phenyltetrazol, 1-phenyltetrazole-thiol
PubChem CID 690730
ChEBI CHEBI:79916
SMILES S=C1N=NNN1C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1265614
View Documents
Thermo Scientific Chemicals
A1265614
25 g
Each for $77.82
 
AAA1265622
View Documents
Thermo Scientific Chemicals
A1265622
100 g
Each for $275.51
 
AAA1265636
View Documents
Thermo Scientific Chemicals
A1265636
500 g
Each for $1,219.26
 
Description

Description

1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1H-tetrazole-5-thiol is an effective inhibitor of aluminum corrosion in 1M HCl solution. 1-Phenyl-1H-tetrazole-5-thiol was used in the synthesis of oxacyclic building blocks via highly stereoselective radical cyclization and olefin metathesis reactions. It was also used in the synthesis of metalated tetradecyl sulfone. It is also used in the spectrophotometric determination of Pt and Bi.

Solubility
Soluble in 5% ethanol, water (partly), acetone, chloroform, and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

86-93-1
178.21
GGZHVNZHFYCSEV-UHFFFAOYSA-N
690730
S=C1N=NNN1C1=CC=CC=C1
C7H6N4S
MFCD00003129
1-phenyl-1h-tetrazole-5-thiol, 1-phenyltetrazole-5-thiol, 1-phenyl-5-mercaptotetrazole, 5-mercapto-1-phenyltetrazole, 1-phenyltetrazoline-5-thione, mercaptophenyltetrazole, phenylmercaptotetrazole, 1-phenyl-5-tetrazolethione, 5-mercapto-1-phenyltetrazol, 1-phenyltetrazole-thiol
CHEBI:79916

Specifications

86-93-1
138°C (280°F)
C7H6N4S
25 g
139068
Soluble in 5% ethanol,water (partly),acetone,chloroform,and methanol.
S=C1N=NNN1C1=CC=CC=C1
178.21
CHEBI:79916
99%
∼145°C (decomposition)
Odorless
MFCD00003129
UN1325
1-phenyl-1h-tetrazole-5-thiol, 1-phenyltetrazole-5-thiol, 1-phenyl-5-mercaptotetrazole, 5-mercapto-1-phenyltetrazole, 1-phenyltetrazoline-5-thione, mercaptophenyltetrazole, phenylmercaptotetrazole, 1-phenyl-5-tetrazolethione, 5-mercapto-1-phenyltetrazol, 1-phenyltetrazole-thiol
GGZHVNZHFYCSEV-UHFFFAOYSA-N
1-phenyl-2H-tetrazole-5-thione
690730
178.22
1-Phenyl-1H-tetrazole-5-thiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P370+P378q-P501c

H228-H317-H319

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 201-710-5

RTECSNumber : XF7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only