missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenylethyl mercaptan, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2634503

 View more versions of this product

Catalog No. AAH2634503


Explore available promotions

Description

Description

The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.

Solubility
Insoluble in water. Solubility in ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications

Specifications

1-Phenylethyl mercaptan
1.021
C8H10S
MFCD00053515
UN2810
1-phenylethyl mercaptan, benzenemethanethiol, .alpha.-methyl, +/--1-phenylethylmercaptan, 1-phenylethane-1-thiol, phenylethanethiol, benzenemethanethiol, alpha-methyl, 1-phenyl ethanethiol, 1-phenyl-ethanethiol, 1-phenethyl mercaptan, 1-phenylethylmercaptan
QZZBJCFNHPYNKO-UHFFFAOYSA-N
1-phenylethanethiol
141850
98%
6263-65-6
76°C (169°F)
1.558
1 g
Air sensitive
Insoluble in water. Solubility in ethanol.
CC(C1=CC=CC=C1)S
138.228
138.23

Chemical Identifiers

6263-65-6
138.228
QZZBJCFNHPYNKO-UHFFFAOYSA-N
141850
CC(C1=CC=CC=C1)S
C8H10S
MFCD00053515
1-phenylethyl mercaptan, benzenemethanethiol, .alpha.-methyl, +/--1-phenylethylmercaptan, 1-phenylethane-1-thiol, phenylethanethiol, benzenemethanethiol, alpha-methyl, 1-phenyl ethanethiol, 1-phenyl-ethanethiol, 1-phenethyl mercaptan, 1-phenylethylmercaptan
1-phenylethanethiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only