Learn More
1-Phenylethyl mercaptan, 98%
CAS: 6263-65-6 | C8H10S | 138.228 g/mol
$90.38 - $448.38
Chemical Identifiers
| CAS | 6263-65-6 |
|---|---|
| Molecular Formula | C8H10S |
| Molecular Weight (g/mol) | 138.228 |
| MDL Number | MFCD00053515 |
| InChI Key | QZZBJCFNHPYNKO-UHFFFAOYSA-N |
| Synonym | 1-phenylethyl mercaptan, benzenemethanethiol, .alpha.-methyl, +/--1-phenylethylmercaptan, 1-phenylethane-1-thiol, phenylethanethiol, benzenemethanethiol, alpha-methyl, 1-phenyl ethanethiol, 1-phenyl-ethanethiol, 1-phenethyl mercaptan, 1-phenylethylmercaptan |
| PubChem CID | 141850 |
| IUPAC Name | 1-phenylethanethiol |
| SMILES | CC(C1=CC=CC=C1)S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2634503
|
Thermo Scientific Chemicals
H2634503 |
1 g |
Each for $90.38
|
|
|||||
|
AAH2634509
|
Thermo Scientific Chemicals
H2634509 |
10 g |
Each for $448.38
|
|
|||||
Description
The compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe compound is used as a source of the thiol functional group in organic synthesis. It is introduced by S-alkylation to give alkyl benzylthioethers.
Solubility
Insoluble in water. Solubility in ethanol.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Chemical Identifiers
| 6263-65-6 | |
| 138.228 | |
| QZZBJCFNHPYNKO-UHFFFAOYSA-N | |
| 141850 | |
| CC(C1=CC=CC=C1)S |
| C8H10S | |
| MFCD00053515 | |
| 1-phenylethyl mercaptan, benzenemethanethiol, .alpha.-methyl, +/--1-phenylethylmercaptan, 1-phenylethane-1-thiol, phenylethanethiol, benzenemethanethiol, alpha-methyl, 1-phenyl ethanethiol, 1-phenyl-ethanethiol, 1-phenethyl mercaptan, 1-phenylethylmercaptan | |
| 1-phenylethanethiol |
Specifications
| 6263-65-6 | |
| 76°C (169°F) | |
| 1.558 | |
| 1 g | |
| Air sensitive | |
| Insoluble in water. Solubility in ethanol. | |
| CC(C1=CC=CC=C1)S | |
| 138.228 | |
| 138.23 | |
| 1-Phenylethyl mercaptan |
| 1.021 | |
| C8H10S | |
| MFCD00053515 | |
| UN2810 | |
| 1-phenylethyl mercaptan, benzenemethanethiol, .alpha.-methyl, +/--1-phenylethylmercaptan, 1-phenylethane-1-thiol, phenylethanethiol, benzenemethanethiol, alpha-methyl, 1-phenyl ethanethiol, 1-phenyl-ethanethiol, 1-phenethyl mercaptan, 1-phenylethylmercaptan | |
| QZZBJCFNHPYNKO-UHFFFAOYSA-N | |
| 1-phenylethanethiol | |
| 141850 | |
| 98% |
Safety and Handling
GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only