missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 426071000

 View more versions of this product

Catalog No. AC426071000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Propynylmagnesium bromide
0.9400g/mL
−1°C
C3H3BrMg
100 mL
1-propynylmagnesium bromide, grignard reagent, bromo prop-1-yn-1-yl magnesium, bromo prop-1-yn-1-yl magnesium, propynylmagnesiumbromide, propynylmagnesiumbromide, prop-1-ynylmagnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, propyn-1-yl magnesium bromide
CC#C[Mg]Br
143.27
143.26
16466-97-0,109-99-9
65.0°C to 67.0°C
AcroSeal™ Glass bottle
MFCD00216588
0.94
YVJBCTFVENNIOV-UHFFFAOYSA-M
magnesium;prop-1-yne;bromide
3608069

Chemical Identifiers

16466-97-0
143.27
YVJBCTFVENNIOV-UHFFFAOYSA-M
3608069
CC#C[Mg]Br
C3H3BrMg
MFCD00216588
1-propynylmagnesium bromide, grignard reagent, bromo prop-1-yn-1-yl magnesium, bromo prop-1-yn-1-yl magnesium, propynylmagnesiumbromide, propynylmagnesiumbromide, prop-1-ynylmagnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, propyn-1-yl magnesium bromide
magnesium;prop-1-yne;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
May form explosive peroxides.
Reacts violentl

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only