Learn More
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$232.24 - $699.15
Chemical Identifiers
CAS | 16466-97-0 |
---|---|
Molecular Formula | C3H3BrMg |
Molecular Weight (g/mol) | 143.27 |
MDL Number | MFCD00216588 |
InChI Key | YVJBCTFVENNIOV-UHFFFAOYSA-M |
Synonym | 1-propynylmagnesium bromide, grignard reagent, 1-propynyl magnesium bromide, 1-propynylmagnesium bromide solution, 1-propynylmagnesium bromide solution, propynyl magnesium bromide, propynyl magnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, propyn-1-yl magnesium bromide |
PubChem CID | 3608069 |
IUPAC Name | magnesium;prop-1-yne;bromide |
SMILES | CC#C[Mg]Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC426071000
|
Thermo Scientific Chemicals
426071000 |
100 mL | AcroSeal™ Glass bottle |
Each for $232.24
|
|
||||
AC426078000
|
Thermo Scientific Chemicals
426078000 |
800 mL | AcroSeal™ Glass bottle |
Each for $699.15
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16466-97-0 | |
143.27 | |
YVJBCTFVENNIOV-UHFFFAOYSA-M | |
3608069 | |
CC#C[Mg]Br |
C3H3BrMg | |
MFCD00216588 | |
1-propynylmagnesium bromide, grignard reagent, 1-propynyl magnesium bromide, 1-propynylmagnesium bromide solution, 1-propynylmagnesium bromide solution, propynyl magnesium bromide, propynyl magnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, propyn-1-yl magnesium bromide | |
magnesium;prop-1-yne;bromide |
Specifications
16466-97-0,109-99-9 | |
65.0°C to 67.0°C | |
AcroSeal™ Glass bottle | |
MFCD00216588 | |
0.94 | |
YVJBCTFVENNIOV-UHFFFAOYSA-M | |
magnesium;prop-1-yne;bromide | |
3608069 | |
1-Propynylmagnesium bromide |
0.9400g/mL | |
−1°C | |
C3H3BrMg | |
100 mL | |
1-propynylmagnesium bromide, grignard reagent, bromo prop-1-yn-1-yl magnesium, bromo prop-1-yn-1-yl magnesium, propynylmagnesiumbromide, propynylmagnesiumbromide, prop-1-ynylmagnesium bromide, prop-1-inyl magnesium bromide, prop-1-ynyl-magnesium bromide, propyn-1-yl magnesium bromide, propyn-1-yl magnesium bromide | |
CC#C[Mg]Br | |
143.27 | |
143.26 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
May form explosive peroxides.
Reacts violentl
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
RUO – Research Use Only