missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Tetralone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1330322

 View more versions of this product

Catalog No. AAA1330322


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Tetralone
6°C to 8°C
257°C
C10H10O
MFCD00001688
607374
XHLHPRDBBAGVEG-UHFFFAOYSA-N
3,4-dihydro-2H-naphthalen-1-one
10724
97%
529-34-0
1.097
129°C (264°F)
1.5695
100 g
alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
O=C1CCCC2=CC=CC=C12
146.19
146.19

Chemical Identifiers

529-34-0
146.19
XHLHPRDBBAGVEG-UHFFFAOYSA-N
10724
O=C1CCCC2=CC=CC=C12
C10H10O
MFCD00001688
alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
3,4-dihydro-2H-naphthalen-1-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P301+P312-P304+P340-P311-P330-P501c

H302-H331

EINECSNumber : 208-460-6

RTECSNumber : QK4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only