missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Tetralone, 97%, Thermo Scientific Chemicals

$109.77 - $351.89

Chemical Identifiers

CAS 529-34-0
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001688
InChI Key XHLHPRDBBAGVEG-UHFFFAOYSA-N
Synonym alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
PubChem CID 10724
IUPAC Name 3,4-dihydro-2H-naphthalen-1-one
SMILES O=C1CCCC2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1330322
View Documents
Thermo Scientific Chemicals
A1330322
100 g
Each for $109.77
 
AAA1330336
View Documents
Thermo Scientific Chemicals
A1330336
500 g
Each for $351.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

529-34-0
146.19
XHLHPRDBBAGVEG-UHFFFAOYSA-N
10724
O=C1CCCC2=CC=CC=C12
C10H10O
MFCD00001688
alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
3,4-dihydro-2H-naphthalen-1-one

Specifications

529-34-0
1.097
129°C (264°F)
1.5695
100 g
alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
O=C1CCCC2=CC=CC=C12
146.19
146.19
1-Tetralone
6°C to 8°C
257°C
C10H10O
MFCD00001688
607374
XHLHPRDBBAGVEG-UHFFFAOYSA-N
3,4-dihydro-2H-naphthalen-1-one
10724
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P301+P312-P304+P340-P311-P330-P501c

H302-H331

EINECSNumber : 208-460-6

RTECSNumber : QK4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only