Learn More
Epredia™ 10% Neutral Buffered Formalin
Reduce exposure to toxic fumes with the premixed Epredia™ Neutral Buffered Formalin (10%). AFIP formulated.
Supplier: Epredia™ 5735
Description
- Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
- Stringent quality control ensures a formalin concentration of 10%
Specifications
| Formalin | |
| 5 Liter |