Learn More
Epredia™ 10% Neutral Buffered Formalin
Reduce exposure to toxic fumes with the premixed Epredia™ Neutral Buffered Formalin (10%). AFIP formulated.
$89.67 - $1533.40
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity | ||||
|
22-050-104
|
Epredia™
SH5701 |
1 gal. | Formalin |
Case of 4 Each for $89.67
|
|
||||
|
22-050-105
|
Epredia™
5705 |
5 gal. | Formalin |
Each for $164.63
|
|
||||
|
57-35
|
Epredia™
5735 |
5 Liter | Formalin |
Case of 4 Each for $380.41
|
|
||||
|
22-110-869
|
Epredia™
SH5725 |
2.5 gal. | Formalin |
Each for $173.77
|
|
||||
|
22-110-614
|
Epredia™
5755 |
55 gal. | Formalin |
Each for $1,533.40
|
|
||||
Description
- Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
- Stringent quality control ensures a formalin concentration of 10%
Specifications
| Formalin |